Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfadiazine sodium | 547-32-0 | 98.0% | 5 G
SDP

Catalog No. 5000406484
Encompass_Preferred

Sulfadiazine sodium is a sulfonamide antibiotic with antimalarial activity, utilized in toxoplasmosis research.

  • Sulfonamide antibiotic with antimalarial activity
  • Useful for toxoplasmosis research
  • Purity of 98.0%
  • Molecular formula: C10H9N4NaO2S
  • Molecular weight: 272.26
  • Appears as a white to off-white solid
  • Store solid at 4°C, protected from light, under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-06484 Supplier Medchemexpress LLC Supplier No. HYB0273A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfadiazine sodium is a sulfonamide antibiotic with antimalarial activity, utilized in toxoplasmosis research.

  • Sulfonamide antibiotic with antimalarial activity
  • Useful for toxoplasmosis research
  • Purity of 98.0%
  • Molecular formula: C10H9N4NaO2S
  • Molecular weight: 272.26
  • Appears as a white to off-white solid
  • Store solid at 4°C, protected from light, under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.