Learn More
Medchemexpress LLC Sulfadiazine sodium | 547-32-0 | 98.0% | 500 MG
SDP
Catalog No. 5000407633
Encompass_Preferred
Sulfadiazine sodium is a sulfonamide antibiotic with antimalarial activity. It is used for toxoplasmosis research, targeting bacteria and parasites in anti-infection pathways.
  • Sulfonamide antibiotic
  • Antimalarial activity
  • Used for toxoplasmosis research
  • Targets bacteria and parasites
  • Involved in anti-infection pathways
Catalog No. 50-004-07633 Supplier Medchemexpress LLC Supplier No. HYB0273A500MG
Sulfadiazine sodium is a sulfonamide antibiotic with antimalarial activity. It is used for toxoplasmosis research, targeting bacteria and parasites in anti-infection pathways.
  • Sulfonamide antibiotic
  • Antimalarial activity
  • Used for toxoplasmosis research
  • Targets bacteria and parasites
  • Involved in anti-infection pathways