Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfadiazine sodium salt, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014573 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4573 25 g
11-101-4574 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4573 Supplier MilliporeSigma Supelco Supplier No. S638725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfadiazine sodium salt is a broad-spectrum antibiotic belonging to the class of sulfonamides commonly used against gram-postive and gram-negative microorganisms in humans and veterinary medication. Its mode of action involves inhibiting the multiplication of bacteria as competitive inhibitors of p-aminobenzoic acid in the folic acid metabolism cycle. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 547-32-0
Physical Form Crystalline Powder
Linear Formula C10H9N4 NaO2 S
MDL Number MFCD00067333
Synonym 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide sodium salt; SPS Agar Supplement
Type Organic Standard
Quantity 25 g
Formula Weight 272.26
Percent Purity ≥98% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.