Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfadiazine USP, Medisca™
GREENER_CHOICESDP

Catalog No. 18048037 Shop All Medisca Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-037 5 kg
18-048-054 25 kg
18-048-094 500 g
18-048-095 1 kg
18-048-117 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-048-037 Supplier Medisca Supplier No. 322403
Add to Cart
Add to Cart

Sulfadiazine USP Not for human use.

Chemical Identifiers

CAS 68-35-9

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Color White to Light Yellow
Quantity 5 kg
Percent Purity 98.0 to 102.0%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.