Learn More
Med Vet International Sulfadimethoxine Injection 40mg/mL, 250mL
Catalog No. 502828918
Encompass_Preferred
For the treatment of bovine respiratory disease complex (shipping fever complex) and bacterial pneumonia associated with Pasteurella spp. sensitive to sulfadimethoxine; necrotic pododermatitis (foot rot) and calf diphtheria caused by Fusobacterium necrophorum sensitive to sulfadimethoxine. NOT FOR SALE IN CALIFORNIA
Catalog No. 50-282-8918 Supplier Med Vet International Supplier No. RXSULFADIMINJ
For the treatment of bovine respiratory disease complex (shipping fever complex) and bacterial pneumonia associated with Pasteurella spp. sensitive to sulfadimethoxine; necrotic pododermatitis (foot rot) and calf diphtheria caused by Fusobacterium necrophorum sensitive to sulfadimethoxine. NOT FOR SALE IN CALIFORNIA