Learn More
Cayman Chemical Sulfadimethoxine, Purity: ≥98%, CAS: 122-11-2, 25 g
SDP
Catalog No. 502940436
Encompass_Preferred
A sulfonamide antibiotic; inhibits recombinant P. carinii DHPS (IC50 = 25 nM) in a cell-free assay; active against S. aureus, S. pneumoniae, K. pneumoniae, and E. coli in a disc assay; active against C. spiroforme isolates from rabbits (MIC90 = 256 μg/ml) and certain M. bovoculi isolates from keratoconjunctivitis infections in cows (MIC50 = ≤256 μg/ml); efficacious S. pneumoniae, K. pneumoniae, and E. coli systemic infections in mice (CD50s = 79.7, 2.7, and 12.2 mg/kg, respectively); efficacious against H. capsulatum systemic fungal infections in mice (CD50 = 300 mg/kg)
Catalog No. 50-294-0436 Supplier Cayman Chemical Supplier No. 2882725 G
A sulfonamide antibiotic; inhibits recombinant P. carinii DHPS (IC50 = 25 nM) in a cell-free assay; active against S. aureus, S. pneumoniae, K. pneumoniae, and E. coli in a disc assay; active against C. spiroforme isolates from rabbits (MIC90 = 256 μg/ml) and certain M. bovoculi isolates from keratoconjunctivitis infections in cows (MIC50 = ≤256 μg/ml); efficacious S. pneumoniae, K. pneumoniae, and E. coli systemic infections in mice (CD50s = 79.7, 2.7, and 12.2 mg/kg, respectively); efficacious against H. capsulatum systemic fungal infections in mice (CD50 = 300 mg/kg)