Learn More
Medchemexpress LLC Sulfadimethoxine | 122-11-2 | 99.1% | 250 MG
SDP
Catalog No. 5000407369
Encompass_Preferred
Sulfadimethoxine (Sulphadimethoxine) is a sulfonamide antibiotic used to treat various infections, including respiratory, urinary tract, enteric, and soft tissue infections. It is also an analytical standard intended for research and analytical applications.
  • Sulfonamide antibiotic for a wide range of infections
  • Effective against respiratory, urinary tract, enteric, and soft tissue infections
  • Analytical standard for research and analysis
  • Purity of 99.10%
  • For research use only
Catalog No. 50-004-07369 Supplier Medchemexpress LLC Supplier No. 250MGHYB0337R
Sulfadimethoxine (Sulphadimethoxine) is a sulfonamide antibiotic used to treat various infections, including respiratory, urinary tract, enteric, and soft tissue infections. It is also an analytical standard intended for research and analytical applications.
  • Sulfonamide antibiotic for a wide range of infections
  • Effective against respiratory, urinary tract, enteric, and soft tissue infections
  • Analytical standard for research and analysis
  • Purity of 99.10%
  • For research use only