Learn More
Apexbio Technology LLC Sulfadimethoxine 122-11-2 50mg
SDP
Catalog No. 502240773
Encompass_Preferred
Sulfadimethoxine (122-11-2) is a small-molecule inhibitor targeting bacterial folic acid synthesis It is designed to inhibit this metabolic pathway thereby suppressing bacterial proliferation Sulfadimethoxine exerts its biological activity primarily through sulfonamide-mediated inhibition of folic acid synthesis in bacteria In immunocompromised rodent models Sulfadimethoxine demonstrates inhibition of Cryptosporidium parvum with an IC50 value of approximately 25 mg/kg in vivo Based on these pharmacological properties Sulfadimethoxine holds research potential in experimental studies of parasitic and bacterial diseases including respiratory tract infections pelvic inflammatory disorders intracranial abscesses and anticryptosporidial applications in veterinary research
This item is not returnable. View return policy
Catalog No. 50-224-0773 Supplier Apexbio Technology LLC Supplier No. B330150
Sulfadimethoxine (122-11-2) is a small-molecule inhibitor targeting bacterial folic acid synthesis It is designed to inhibit this metabolic pathway thereby suppressing bacterial proliferation Sulfadimethoxine exerts its biological activity primarily through sulfonamide-mediated inhibition of folic acid synthesis in bacteria In immunocompromised rodent models Sulfadimethoxine demonstrates inhibition of Cryptosporidium parvum with an IC50 value of approximately 25 mg/kg in vivo Based on these pharmacological properties Sulfadimethoxine holds research potential in experimental studies of parasitic and bacterial diseases including respiratory tract infections pelvic inflammatory disorders intracranial abscesses and anticryptosporidial applications in veterinary research