Learn More
Medchemexpress LLC Sulfadimethoxine-d6 | 73068-02-7 | 99.0% | 1 MG
SDP
Catalog No. 5000405311
Encompass_Preferred
Sulfadimethoxine-d6 is a deuterium-labeled version of Sulfadimethoxine, a sulfonamide antibiotic used to treat various infections. It is a stable isotope-labeled compound primarily used as a tracer and internal standard in quantitative analysis.
  • Deuterium-labeled compound
  • Used as a tracer
  • Internal standard for NMR, GC-MS, or LC-MS analysis
  • Intended for research use only
  • Stable under recommended storage conditions
  • Not classified as a hazardous substance or mixture
Catalog No. 50-004-05311 Supplier Medchemexpress LLC Supplier No. HYB0337S11MG
Sulfadimethoxine-d6 is a deuterium-labeled version of Sulfadimethoxine, a sulfonamide antibiotic used to treat various infections. It is a stable isotope-labeled compound primarily used as a tracer and internal standard in quantitative analysis.
  • Deuterium-labeled compound
  • Used as a tracer
  • Internal standard for NMR, GC-MS, or LC-MS analysis
  • Intended for research use only
  • Stable under recommended storage conditions
  • Not classified as a hazardous substance or mixture