Learn More
Medchemexpress LLC Sulfadimethoxine (standard) | 122-11-2 | 99.86% | 100 MG
SDP
Catalog No. 5000406280
Encompass_Preferred
Sulfadimethoxine (Standard) is the analytical standard of Sulfadimethoxine. This product is intended for research and analytical applications. Sulfadimethoxine (Sulphadimethoxine) is a sulfonamide antibiotic used to treat many infections.
  • Analytical standard for research and analytical applications
  • Reference standard supplied assay
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-06280 Supplier Medchemexpress LLC Supplier No. HYB0337R100MG
Sulfadimethoxine (Standard) is the analytical standard of Sulfadimethoxine. This product is intended for research and analytical applications. Sulfadimethoxine (Sulphadimethoxine) is a sulfonamide antibiotic used to treat many infections.
  • Analytical standard for research and analytical applications
  • Reference standard supplied assay
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS