Learn More
Medchemexpress LLC Sulfadimethoxine (Standard) | 122-11-2 | 99.86% | 50 MG
SDP
Catalog No. 5000406204
Encompass_Preferred
Sulfadimethoxine (Standard) is the analytical standard of Sulfadimethoxine, a sulfonamide antibiotic used to treat various infections. This product is intended solely for research and analytical applications.
  • Serves as a reference standard for assays.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Applicable for techniques such as HPLC, GC, and MS.
  • For research use only.
Catalog No. 50-004-06204 Supplier Medchemexpress LLC Supplier No. HYB0337R50MG
Sulfadimethoxine (Standard) is the analytical standard of Sulfadimethoxine, a sulfonamide antibiotic used to treat various infections. This product is intended solely for research and analytical applications.
  • Serves as a reference standard for assays.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Applicable for techniques such as HPLC, GC, and MS.
  • For research use only.