Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfadimethoxine, Thermo Scientific™

Catalog No. AC469761000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469761000 100 g
AC469760250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469761000 Supplier Thermo Scientific Chemicals Supplier No. 469761000
Only null left
Add to Cart
This item is not returnable. View return policy

Specifications

Type Sulfadimethoxine
Chemical Name or Material Sulfadimethoxine
Physical Form Crystalline Powder
CAS 122-11-2
Melting Point 195°C to 203°C
Infrared Spectrum Conforms
Loss on Drying ≤0.5 % (1 g, 105°C)
Packaging Glass Bottle
Color Cream to White
Quantity 100 g
Molecular Formula C12 H14 N4 O4 S
Product Line Life science
MDL Number 00057345
Shipping Condition No special requirements
Molecular Weight (g/mol) 310.33
Percent Purity ≥97.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.