Learn More
Cayman Chemical SulfadoxIn-d4 5mg
SDP
Catalog No. 502915427
Encompass_Preferred
An internal standard for the quantification of sulfadoxin by GC- or LC-MS
Catalog No. 50-291-5427 Supplier Cayman Chemical Supplier No. 372005 MG
An internal standard for the quantification of sulfadoxin by GC- or LC-MS