Learn More
Medchemexpress LLC HY-B0439 500mg Medchemexpress, Sulfadoxine CAS:2447-57-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0439500MG
Encompass_Preferred
Medchemexpress, HY-B0439 500mg Sulfadoxine CAS:2447-57-6 Sulfadoxine(Sulphadoxine) is a long acting sulfonamide that is used, usually in combination with other drugs, for respiratory, urinary tract and malarial infections. Sulfadoxine inhibits HIV replication in peripheral blood mononuclear cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2539