Learn More
Apexbio Technology LLC Sulfadoxine 2447-57-6 5g
SDP
Catalog No. 502240753
Encompass_Preferred
Sulfadoxine (CAS 2447-57-6) is a small-molecule inhibitor targeting dihydropteroate synthase It is designed to competitively inhibit this enzyme thereby disrupting folate biosynthesis in Plasmodium parasites Sulfadoxine exerts its biological activity primarily through inhibition of dihydropteroate synthase mediated folate synthesis In cell-based studies sulfadoxine demonstrates inhibitory activity against human immunodeficiency virus (HIV) replication within peripheral blood mononuclear cells (PBMCs) with an IC50 value in the micromolar range Based on these pharmacological properties sulfadoxine holds research potential in antimalarial drug investigation parasite metabolic pathway studies pathogen susceptibility assays antiviral pharmacological experiments and investigations of host-pathogen interactions and viral replication mechanisms
This item is not returnable. View return policy
Catalog No. 50-224-0753 Supplier Apexbio Technology LLC Supplier No. B18415000
Sulfadoxine (CAS 2447-57-6) is a small-molecule inhibitor targeting dihydropteroate synthase It is designed to competitively inhibit this enzyme thereby disrupting folate biosynthesis in Plasmodium parasites Sulfadoxine exerts its biological activity primarily through inhibition of dihydropteroate synthase mediated folate synthesis In cell-based studies sulfadoxine demonstrates inhibitory activity against human immunodeficiency virus (HIV) replication within peripheral blood mononuclear cells (PBMCs) with an IC50 value in the micromolar range Based on these pharmacological properties sulfadoxine holds research potential in antimalarial drug investigation parasite metabolic pathway studies pathogen susceptibility assays antiviral pharmacological experiments and investigations of host-pathogen interactions and viral replication mechanisms