Learn More
Medchemexpress LLC Sulfadoxine-d3 | 1262770-70-6 | MFCD10567357 | 98.0% | 313.35 | C12H11D3N4O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0439S110MG
Encompass_Preferred
Sulfadoxine-d3 is the deuterium-labeled isotopologue of sulfadoxine, a long-acting sulfonamide anti-malarial agent provided as a high-purity analytical standard for research. It is used as an internal standard, tracer, or reference material in pharmacokinetic and mass spectrometry studies.
  • Deuterium-labeled analog for tracer and internal standard use.
  • High purity, 98.0%, suitable for analytical applications.
  • Soluble in DMSO, limited solubility in water.
  • Stable as a powder when stored at -20°C, with defined in-solution storage limits.
  • Supplied in small analytical quantities useful for method development and validation.
Catalog No. 50-002-98911