Learn More
Medchemexpress LLC Sulfadoxine (Standard) | 2447-57-6 | 98.8% | 250 MG
SDP
Catalog No. 5000406811
Encompass_Preferred
Sulfadoxine (Standard) is the analytical standard of Sulfadoxine, intended for research and analytical applications. This long-acting sulfonamide is used, often in combination with other agents, for respiratory, urinary tract, and malarial infections. It also inhibits HIV replication in peripheral blood mononuclear cells.
  • Analytical standard grade
  • Reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
Catalog No. 50-004-06811 Supplier Medchemexpress LLC Supplier No. 250MGHYB0439R
Sulfadoxine (Standard) is the analytical standard of Sulfadoxine, intended for research and analytical applications. This long-acting sulfonamide is used, often in combination with other agents, for respiratory, urinary tract, and malarial infections. It also inhibits HIV replication in peripheral blood mononuclear cells.
  • Analytical standard grade
  • Reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS