Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfadoxine (Sulphadoxine), G-Biosciences
SCI_ED

Catalog No. S000016
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000016 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000016 Supplier G-Biosciences Supplier No. RC1106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfadoxine is a broad-spectrum sulfonamide antibiotic effective against gram-negative and gram-positive bacteria.

It is commonly used against malaria, especially Plasmodium falciparum malaria with chloroquine resistance. It has been used to study Toxoplasma gondii infections and to prevent infections in post-surgical animal models.

Chemical Identifiers

Molecular Weight (g/mol) 310.33

Specifications

Physical Form Powder
Quantity 10 g
Product Line Gibco
Boiling Point 522.8°C
Percent Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.