Learn More
Cayman Chemical SulfaethoxypyrIdazIn 10mg
SDP
Catalog No. 502942851
Encompass_Preferred
A sulfonamide antibiotic; active against several strains of E. coli isolated from necropsied animals or poultry at 0.25 or 1 mg/ml; induces the formation of cataracts in dogs when administered at 150 mg/kg for 13-27 weeks
Catalog No. 50-294-2851 Supplier Cayman Chemical Supplier No. 4169810 MG
A sulfonamide antibiotic; active against several strains of E. coli isolated from necropsied animals or poultry at 0.25 or 1 mg/ml; induces the formation of cataracts in dogs when administered at 150 mg/kg for 13-27 weeks