Learn More
Cayman Chemical SulfaethoxypyrIdazIn 50mg
SDP
Catalog No. 502943840
Encompass_Preferred
A sulfonamide antibiotic; active against several strains of E. coli isolated from necropsied animals or poultry at 0.25 or 1 mg/ml; induces the formation of cataracts in dogs when administered at 150 mg/kg for 13-27 weeks
Catalog No. 50-294-3840 Supplier Cayman Chemical Supplier No. 4169850 MG
A sulfonamide antibiotic; active against several strains of E. coli isolated from necropsied animals or poultry at 0.25 or 1 mg/ml; induces the formation of cataracts in dogs when administered at 150 mg/kg for 13-27 weeks