Learn More
Medchemexpress LLC HY-B1267 500mg Medchemexpress, Sulfaguanidine CAS:57-67-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1267500MG
Encompass_Preferred
Medchemexpress, HY-B1267 500mg Sulfaguanidine CAS:57-67-0 Sulfaguanidine is an orally active antimicrobial agent/antibiotic of sulfonamide class. Sulfaguanidine can be used for the research of enteric infections such as bacillary dysentery. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2695