Learn More
Apexbio Technology LLC Sulfaguanidine 57-67-0 50mg
SDP
Catalog No. 502240713
Encompass_Preferred
Sulfaguanidine (57-67-0) is a small-molecule inhibitor targeting bacterial dihydropteroate synthase It is designed to competitively inhibit this enzyme thereby interfering with folic acid biosynthesis and disrupting a pathway critical for bacterial growth Sulfaguanidine exerts its biological activity primarily through competitive inhibition of bacterial dihydropteroate synthase In in vitro infection models sulfaguanidine demonstrates antimicrobial activity with reported IC50 values generally in the micromolar range depending on bacterial species experimental conditions and assay methodologies Based on these pharmacological properties sulfaguanidine holds research potential in studies of antimicrobial resistance mechanisms bacterial folate metabolism and interactions with other antimicrobial agents
This item is not returnable. View return policy
Catalog No. 50-224-0713 Supplier Apexbio Technology LLC Supplier No. B184250
Sulfaguanidine (57-67-0) is a small-molecule inhibitor targeting bacterial dihydropteroate synthase It is designed to competitively inhibit this enzyme thereby interfering with folic acid biosynthesis and disrupting a pathway critical for bacterial growth Sulfaguanidine exerts its biological activity primarily through competitive inhibition of bacterial dihydropteroate synthase In in vitro infection models sulfaguanidine demonstrates antimicrobial activity with reported IC50 values generally in the micromolar range depending on bacterial species experimental conditions and assay methodologies Based on these pharmacological properties sulfaguanidine holds research potential in studies of antimicrobial resistance mechanisms bacterial folate metabolism and interactions with other antimicrobial agents