Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfaguanidine | 57-67-0 | MFCD00038136 | 99.4% | 214.24 g/mol | C7H10N4O2S | 10 MG
SDP

Catalog No. 5000298218
Encompass_Preferred

Sulfaguanidine (Standard) is an analytical reference standard of the sulfonamide-class antibiotic sulfaguanidine provided as a 10 mg vial for research and analytical applications. It is intended for chromatographic, spectrometric, and method validation workflows that require authenticated reference material.

  • Analytical reference material supplied as a 10 mg vial.
  • High purity (99.42%) suitable for analytical assays.
  • Molecular formula C7H10N4O2S; molecular weight 214.24 g/mol.
  • Intended for chromatographic, spectrometric, and method validation workflows.
  • Batch-specific information provided for traceability.

Catalog No. 50-002-98218 Supplier Medchemexpress LLC Supplier No. HYB1267R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfaguanidine (Standard) is an analytical reference standard of the sulfonamide-class antibiotic sulfaguanidine provided as a 10 mg vial for research and analytical applications. It is intended for chromatographic, spectrometric, and method validation workflows that require authenticated reference material.

  • Analytical reference material supplied as a 10 mg vial.
  • High purity (99.42%) suitable for analytical assays.
  • Molecular formula C7H10N4O2S; molecular weight 214.24 g/mol.
  • Intended for chromatographic, spectrometric, and method validation workflows.
  • Batch-specific information provided for traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.