Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfaguanidine, 98%, Thermo Scientific Chemicals

Catalog No. AC431305000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431305000 500 g Glass bottle
AC431300250 25 g Glass bottle
AC431301000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC431305000 Supplier Thermo Scientific Chemicals Supplier No. 431305000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-67-0
Molecular Formula C7H10N4O2S
Molecular Weight (g/mol) 214.25
MDL Number MFCD00038136
InChI Key BRBKOPJOKNSWSG-UHFFFAOYSA-N
Synonym sulfaguanidine, sulphaguanidine, sulfaguanidin, guanicil, sulfaguine, aterian, sulfanilguanidine, sulfanilylguanidine, sulfoguanidine, abiguanil
PubChem CID 5324
IUPAC Name 2-(4-aminophenyl)sulfonylguanidine
SMILES C1=CC(=CC=C1N)S(=O)(=O)N=C(N)N

Specifications

Melting Point 190.5°C to 192.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 97.5% min. (HPLC)
Merck Index 15,904
Formula Weight 214.25
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Sulfaguanidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 200-345-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.