Learn More
Medchemexpress LLC Sulfalene-13C6 | 99.0% | C513C6H12N4O3S | 1 MG
SDP
Catalog No. 5000432940
Encompass_Preferred
Sulfalene-13C6 is the 13C6 labeled version of Sulfalene, an antimalarial agent and long-acting sulfonamide antibacterial. Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules for quantitation during drug development, potentially influencing pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Internal standard for quantitative analysis
  • Applicable in metabolic flux analysis
  • Useful for therapeutic drug monitoring
Catalog No. 50-004-32940 Supplier Medchemexpress LLC Supplier No. HYA0130S1MG
Sulfalene-13C6 is the 13C6 labeled version of Sulfalene, an antimalarial agent and long-acting sulfonamide antibacterial. Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules for quantitation during drug development, potentially influencing pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Internal standard for quantitative analysis
  • Applicable in metabolic flux analysis
  • Useful for therapeutic drug monitoring