Learn More
Medchemexpress LLC HY-A0130 50mg Medchemexpress, Sulfalene CAS:152-47-6 Purity:>98%
SDP
Catalog No. 502082328
Encompass_Preferred
Medchemexpress, HY-A0130 50mg Sulfalene CAS:152-47-6 Sulfalene (Sulfametopyrazine) is an antimalarial agent. Sulfalene is also a long-acting sulfonamide antibacterial. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2328 Supplier Medchemexpress LLC Supplier No. HYA013050MG
Medchemexpress, HY-A0130 50mg Sulfalene CAS:152-47-6 Sulfalene (Sulfametopyrazine) is an antimalarial agent. Sulfalene is also a long-acting sulfonamide antibacterial. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.