Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfamerazine | 127-79-7 | 99.8% | 1 G
SDP

Catalog No. 5000405980
Encompass_Preferred

Sulfamerazine (RP-2632) is a sulfonamide antibacterial, identified as the monomethyl derivative of sulfadiazine (2-sulfanilamido-4-methylpyrimidine). It acts as a bacteriostatic agent by inhibiting bacterial synthesis of dihydrofolic acid, competing with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthesizes, which ultimately reduces the synthesis of bacterial nucleotides and DNA.

  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 264.30
  • Formula: C11H12N4O2S
  • Solubility in DMSO: ≥ 100 mg/mL (378.36 mM)
  • Solubility in H2O: < 0.1 mg/mL (insoluble)

Catalog No. 50-004-05980 Supplier Medchemexpress LLC Supplier No. HYB05121G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfamerazine (RP-2632) is a sulfonamide antibacterial, identified as the monomethyl derivative of sulfadiazine (2-sulfanilamido-4-methylpyrimidine). It acts as a bacteriostatic agent by inhibiting bacterial synthesis of dihydrofolic acid, competing with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthesizes, which ultimately reduces the synthesis of bacterial nucleotides and DNA.

  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 264.30
  • Formula: C11H12N4O2S
  • Solubility in DMSO: ≥ 100 mg/mL (378.36 mM)
  • Solubility in H2O: < 0.1 mg/mL (insoluble)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.