Learn More
Medchemexpress LLC Sulfamerazine | 127-79-7 | 99.8% | 5 G
SDP
Catalog No. 5000407560
Encompass_Preferred
Sulfamerazine (RP-2632) is a sulfonamide antibacterial. It is a monomethyl derivative of sulfadiazine, specifically 2-sulfanilamido-4-methylpyrimidine. This sulfonamide drug inhibits bacterial synthesis of dihydrofolic acid by competing with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthesizes. Sulfamerazine is bacteriostatic in nature; its inhibition of dihydrofolic acid synthesis reduces the synthesis of bacterial nucleotides and DNA.
  • Sulfonamide antibacterial.
  • Monomethyl derivative of sulfadiazine.
  • Inhibits bacterial synthesis of dihydrofolic acid.
  • Competes with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthesizes.
  • Bacteriostatic in nature.
  • Reduces synthesis of bacterial nucleotides and DNA.
Catalog No. 50-004-07560 Supplier Medchemexpress LLC Supplier No. HYB05125G
Sulfamerazine (RP-2632) is a sulfonamide antibacterial. It is a monomethyl derivative of sulfadiazine, specifically 2-sulfanilamido-4-methylpyrimidine. This sulfonamide drug inhibits bacterial synthesis of dihydrofolic acid by competing with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthesizes. Sulfamerazine is bacteriostatic in nature; its inhibition of dihydrofolic acid synthesis reduces the synthesis of bacterial nucleotides and DNA.
  • Sulfonamide antibacterial.
  • Monomethyl derivative of sulfadiazine.
  • Inhibits bacterial synthesis of dihydrofolic acid.
  • Competes with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthesizes.
  • Bacteriostatic in nature.
  • Reduces synthesis of bacterial nucleotides and DNA.