Learn More
Medchemexpress LLC Sulfamerazine-d4 | 1020719-84-9 | MFCD03425631 | 99.0% | 1 MG
SDP
Catalog No. 5000407638
Encompass_Preferred
Sulfamerazine-d4 is a deuterium-labeled version of Sulfamerazine. Sulfamerazine is a sulfonamide antibacterial that works by inhibiting bacterial synthesis of dihydrofolic acid. It does this by competing with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthesizes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07638 Supplier Medchemexpress LLC Supplier No. HYB0512S1MG
Sulfamerazine-d4 is a deuterium-labeled version of Sulfamerazine. Sulfamerazine is a sulfonamide antibacterial that works by inhibiting bacterial synthesis of dihydrofolic acid. It does this by competing with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthesizes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.