Learn More
Medchemexpress LLC Sulfamerazine-d4 | 1020719-84-9 | MFCD03425631 | 99.00% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0512S5MG
Encompass_Preferred
Sulfamerazine-d4 is a deuterium-labeled variant of Sulfamerazine, which is classified as a sulfonamide antibacterial. Its mechanism of action involves inhibiting the bacterial synthesis of dihydrofolic acid. This is achieved by competing with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthesizes.
  • Deuterium labeled
  • Internal standard
  • Antibacterial properties
Catalog No. 50-004-07925