Learn More
Cayman Chemical Sulfamerazine (sodium salt), Purity: ≥98%, CAS: 127-58-2, 50 g
SDP
Catalog No. 502993882
Encompass_Preferred
A sulfonamide antibiotic; inhibits DHPS (IC50 = 11.26 ng/ml for the S. pneumoniae enzyme) and bacterial carbonic anhydrase by ~20% at 500 μM; reduces severity of peribronchitis and inhibits cilia-associated respiratory Bacillus colonization in mice at 500 mg/L; prevents death of Toxoplasma-infected mice
Catalog No. 50-299-3882 Supplier Cayman Chemical Supplier No. 2338050 G
A sulfonamide antibiotic; inhibits DHPS (IC50 = 11.26 ng/ml for the S. pneumoniae enzyme) and bacterial carbonic anhydrase by ~20% at 500 μM; reduces severity of peribronchitis and inhibits cilia-associated respiratory Bacillus colonization in mice at 500 mg/L; prevents death of Toxoplasma-infected mice