Learn More
Medchemexpress LLC HY-B0213 500mg Medchemexpress, Sulfameter CAS:651-06-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0213500MG
Encompass_Preferred
Medchemexpress, HY-B0213 500mg Sulfameter CAS:651-06-9 Sulfameter (Sulfametoxydiazine; 5-Methoxysulfadiazine) is an effective long-acting sulfonamide antibiotic with antibacterial activities. Sulfameter can be used for the research of urinary tract infections and lepriasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2412