Learn More
Cayman Chemical Sulfamethazine, Purity: ≥98%, CAS: 57-68-1, 50 g
SDP
Catalog No. 502984985
Encompass_Preferred
A sulfonamide antibiotic; inhibits DHPS (IC50 = 5.7 µM for the T. gondii enzyme); active against A. pleuropneumoniae (MIC = 32 µg/ml) and enhances the antibacterial activity of trimethoprim against E. coli; has been detected in environmental water samples
Catalog No. 50-298-4985 Supplier Cayman Chemical Supplier No. 2097650 G
A sulfonamide antibiotic; inhibits DHPS (IC50 = 5.7 µM for the T. gondii enzyme); active against A. pleuropneumoniae (MIC = 32 µg/ml) and enhances the antibacterial activity of trimethoprim against E. coli; has been detected in environmental water samples