Learn More
Medchemexpress LLC Sulfamethazine-13C6 | 77643-91-5 | 99.1% | C613C6H14N4O2S | 1 MG
SDP
Catalog No. 5000432495
Encompass_Preferred
Sulfamethazine-13C6 is a 13C-labeled compound and an antimicrobial used to treat and prevent various animal diseases, such as gastrointestinal and respiratory tract infections. It can serve as a tracer and an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Useful for metabolic flux analysis (MFA)
  • Suitable for therapeutic drug monitoring (TDM)
Catalog No. 50-004-32495 Supplier Medchemexpress LLC Supplier No. HYB0035S21MG
Sulfamethazine-13C6 is a 13C-labeled compound and an antimicrobial used to treat and prevent various animal diseases, such as gastrointestinal and respiratory tract infections. It can serve as a tracer and an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Useful for metabolic flux analysis (MFA)
  • Suitable for therapeutic drug monitoring (TDM)