Learn More
Medchemexpress LLC Sulfamethazine-d4 | 1020719-82-7 | 99.9% | C12H10D4N4O2S | 1 MG
SDP
Catalog No. 5000433409
Encompass_Preferred
Sulfamethazine-d4 (Sulfadimidine-d4) is a deuterium labeled Sulfamethazine (Sulfadimidine). Sulfamethazine is an antimicrobial that is widely used to treat and prevent various animal diseases (such as gastrointestinal and respiratory tract infections). This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
  • Antimicrobial used to treat and prevent animal gastrointestinal and respiratory tract infections.
Catalog No. 50-004-33409 Supplier Medchemexpress LLC Supplier No. HYB0035S1MG
Sulfamethazine-d4 (Sulfadimidine-d4) is a deuterium labeled Sulfamethazine (Sulfadimidine). Sulfamethazine is an antimicrobial that is widely used to treat and prevent various animal diseases (such as gastrointestinal and respiratory tract infections). This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
  • Antimicrobial used to treat and prevent animal gastrointestinal and respiratory tract infections.