Learn More
Medchemexpress LLC Sulfamethazine-d4 | 1020719-82-7 | 99.9% | C12H10D4N4O2S | 5 MG
SDP
Catalog No. 5000432193
Encompass_Preferred
Sulfamethazine-d4 is a deuterium-labeled version of Sulfamethazine, an antimicrobial used to treat and prevent various animal diseases, including gastrointestinal and respiratory tract infections. This compound is suitable for use as a tracer and internal standard for quantitative analysis in techniques such as NMR, GC-MS, or LC-MS. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for analytical methods
  • Impacts pharmacokinetic and metabolic profiles of drugs
  • Suitable for research applications
Catalog No. 50-004-32193 Supplier Medchemexpress LLC Supplier No. HYB0035S5MG
Sulfamethazine-d4 is a deuterium-labeled version of Sulfamethazine, an antimicrobial used to treat and prevent various animal diseases, including gastrointestinal and respiratory tract infections. This compound is suitable for use as a tracer and internal standard for quantitative analysis in techniques such as NMR, GC-MS, or LC-MS. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for analytical methods
  • Impacts pharmacokinetic and metabolic profiles of drugs
  • Suitable for research applications