Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamethazine, G-Biosciences
SCI_ED

Catalog No. S139827
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139827 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139827 Supplier G-Biosciences Supplier No. RC1107
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfamethazine is broad-spectrum sulfonamide antibiotic targeting bacteria causing bronchitis and urinary tract infections.

It is used in disposition and depletion studies of kinetics and to develop procedures used for quantification. Sulfamethazine is also an inducer of CYP3A4.

Chemical Identifiers

Molecular Weight (g/mol) 278.33

Specifications

Physical Form Powder
Quantity 100 g
Product Line Gibco
Boiling Point 420.4°C
Percent Purity >98%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.