Learn More
Medchemexpress LLC Sulfamethazine (sodium) | 1981-58-4 | 99.9% | C12H13N4NaO2S | 5 G
Supplier: Medchemexpress LLC HYB0035A5G
Sulfamethazine (sodium) is an antimicrobial with blood-brain barrier permeability. It is widely used to treat and prevent various animal diseases, such as gastrointestinal and respiratory tract infections. This product is for research use only.
- Antimicrobial with blood-brain barrier permeability
- Treats animal gastrointestinal and respiratory tract infections
- Maximum residue level in animal products is 100 μg/kg (China and European Commission)
- For research use only