Learn More
Selleck Chemical LLC Sulfamethazine Sodium Salt S5210-25mg
SDP
Catalog No. 503112264
Encompass_Preferred
Sulfamethazine (Sulfadimethyldiazine) is a sulfanilamide anti-infective agent used in the lifestock industry
Catalog No. 50-311-2264 Supplier Selleck Chemical LLC Supplier No. S521025MG
Sulfamethazine (Sulfadimethyldiazine) is a sulfanilamide anti-infective agent used in the lifestock industry