Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamethazine sodium salt, MP Biomedicals™

Catalog No. ICN15670980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15670980 100 g
ICN15670983 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15670980 Supplier MP Biomedicals, Inc Supplier No. 0215670980
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1981-58-4
Molecular Formula C12H13N4NaO2S
Molecular Weight (g/mol) 300.312
InChI Key NGIVTUVVBWOTNT-UHFFFAOYSA-N
Synonym sulfamethazine sodium salt, sulfamethazine sodium, sulfadimidine sodium, sodium sulfamethazine, sodium sulfametazine, sodium sulfamethamide, sodium sulfamezathine, bovibol, vesadin, sodium sulfamethiazine
PubChem CID 13456556
IUPAC Name sodium;(4-aminophenyl)sulfonyl-(4,6-dimethylpyrimidin-2-yl)azanide
SMILES CC1=CC(=NC(=N1)[N-]S(=O)(=O)C2=CC=C(C=C2)N)C.[Na+]

Specifications

Quantity 100 g
Formula Weight 300.3
Physical Form Solid
Chemical Name or Material Sulfamethazine sodium salt
EINECSNumber 217-840-0
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.