Learn More
Medchemexpress LLC Sulfamethazine (Standard) | 57-68-1 | 99.5% | C₁₂H₁₄N₄O₂S | 100 MG
SDP
Catalog No. 5000432494
Encompass_Preferred
Sulfamethazine (Standard) is an analytical standard of Sulfamethazine intended for research and analytical applications. It is an antimicrobial widely used to treat and prevent various animal diseases, including gastrointestinal and respiratory tract infections.
  • Analytical standard for Sulfamethazine
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Can be used to treat and prevent animal diseases
  • Reference standard supplied assay
Catalog No. 50-004-32494 Supplier Medchemexpress LLC Supplier No. HYB0035R100MG
Sulfamethazine (Standard) is an analytical standard of Sulfamethazine intended for research and analytical applications. It is an antimicrobial widely used to treat and prevent various animal diseases, including gastrointestinal and respiratory tract infections.
  • Analytical standard for Sulfamethazine
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Can be used to treat and prevent animal diseases
  • Reference standard supplied assay