Learn More
Medchemexpress LLC Sulfamethazine (standard) | 57-68-1 | MFCD00006066 | 99.5% | C₁₂H₁₄N₄O₂S | 50 MG
SDP
Catalog No. 5000433428
Encompass_Preferred
Sulfamethazine (Standard) is the analytical standard of Sulfamethazine. It is an antimicrobial used to treat and prevent various animal diseases, including gastrointestinal and respiratory tract infections. This product is intended for research and analytical applications, and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard
  • Antimicrobial for animal disease treatment and prevention
  • Used in gastrointestinal and respiratory tract infection research
  • Suitable for HPLC, GC, and MS experiments
  • Reference standard for assay
Catalog No. 50-004-33428 Supplier Medchemexpress LLC Supplier No. HYB0035R50MG
Sulfamethazine (Standard) is the analytical standard of Sulfamethazine. It is an antimicrobial used to treat and prevent various animal diseases, including gastrointestinal and respiratory tract infections. This product is intended for research and analytical applications, and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard
  • Antimicrobial for animal disease treatment and prevention
  • Used in gastrointestinal and respiratory tract infection research
  • Suitable for HPLC, GC, and MS experiments
  • Reference standard for assay