Learn More
Medchemexpress LLC Sulfamethazine (Standard) | 57-68-1 | MFCD00006066 | 99.5% | C12H14N4O2S | 25 MG
SDP
Catalog No. 5000359160
Encompass_Preferred
Sulfamethazine (Standard) is an analytical standard intended for research and analytical applications. This antimicrobial, also known as Sulfadimidine, is widely used to treat and prevent various animal diseases, including gastrointestinal and respiratory tract infections. Regulatory bodies like China and the European Commission set the maximum residue level for Sulfamethazine in animal products at 100 μg/kg.
  • Used as an analytical reference standard
  • Applicable in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Antimicrobial properties for treating animal diseases
Catalog No. 50-003-59160 Supplier Medchemexpress LLC Supplier No. 25MGHYB0035R
Sulfamethazine (Standard) is an analytical standard intended for research and analytical applications. This antimicrobial, also known as Sulfadimidine, is widely used to treat and prevent various animal diseases, including gastrointestinal and respiratory tract infections. Regulatory bodies like China and the European Commission set the maximum residue level for Sulfamethazine in animal products at 100 μg/kg.
  • Used as an analytical reference standard
  • Applicable in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Antimicrobial properties for treating animal diseases