Learn More
Medchemexpress LLC Sulfamethizole | 144-82-1 | MFCD00053363 | 99.8% | 50 MG
SDP
Catalog No. 5000405512
Encompass_Preferred
Sulfamethizole (Standard) is an analytical standard and a sulfathiazole antibiotic. It functions by inhibiting the synthesis of folic acid and thymine. This compound selectively suppresses Photobacterium phosphoreum bioluminescence and demonstrates antibacterial activity against Escherichia coli-mediated urinary tract infections. It is intended for research and analytical applications, and is suitable for qualitative, quantitative, and methodological research experiments in techniques like HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Sulfathiazole antibiotic
  • Inhibits folic acid and thymine synthesis
  • Selectively suppresses Photobacterium phosphoreum bioluminescence
  • Active against Escherichia coli-mediated urinary tract infections
  • Suitable for research on urinary tract infections and bioluminescence mechanisms
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-004-05512 Supplier Medchemexpress LLC Supplier No. HYB0333R50MG
Sulfamethizole (Standard) is an analytical standard and a sulfathiazole antibiotic. It functions by inhibiting the synthesis of folic acid and thymine. This compound selectively suppresses Photobacterium phosphoreum bioluminescence and demonstrates antibacterial activity against Escherichia coli-mediated urinary tract infections. It is intended for research and analytical applications, and is suitable for qualitative, quantitative, and methodological research experiments in techniques like HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Sulfathiazole antibiotic
  • Inhibits folic acid and thymine synthesis
  • Selectively suppresses Photobacterium phosphoreum bioluminescence
  • Active against Escherichia coli-mediated urinary tract infections
  • Suitable for research on urinary tract infections and bioluminescence mechanisms
  • Used in HPLC, GC, and MS research experiments