Learn More
Cayman Chemical Sulfamethizole, Purity: ≥98%, CAS: 144-82-1, 250 mg
SDP
Catalog No. 502993894
Encompass_Preferred
A sulfonamide antibiotic (MIC90s = 1.25-5,000 μg/ml against clinical isolates of E. coli and K. pneumoniae); inhibits dihydropteroate synthase, an enzyme involved in folate biosynthesis
Catalog No. 50-299-3894 Supplier Cayman Chemical Supplier No. 23355250 MG
A sulfonamide antibiotic (MIC90s = 1.25-5,000 μg/ml against clinical isolates of E. coli and K. pneumoniae); inhibits dihydropteroate synthase, an enzyme involved in folate biosynthesis