Learn More
Cayman Chemical Sulfamethizole-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2470130-12-0, 1 mg
SDP
Catalog No. 502935169
Encompass_Preferred
An internal standard for the quantification of sulfamethizole by GC- or LC-MS
Catalog No. 50-293-5169 Supplier Cayman Chemical Supplier No. 267891 MG
An internal standard for the quantification of sulfamethizole by GC- or LC-MS