Learn More
Cayman Chemical Sulfamethizole-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2470130-12-0, 5 mg
SDP
Catalog No. 502932670
Encompass_Preferred
An internal standard for the quantification of sulfamethizole by GC- or LC-MS
Catalog No. 50-293-2670 Supplier Cayman Chemical Supplier No. 267895 MG
An internal standard for the quantification of sulfamethizole by GC- or LC-MS