Learn More
Medchemexpress LLC Sulfamethizole (standard) | 144-82-1 | 99.79% | 25 MG
SDP
Catalog No. 5000450573
Encompass_Preferred
Sulfamethizole (Standard) is the analytical standard of Sulfamethizole (HY-B0333). It is intended for research and analytical applications. Sulfamethizole is a sulfathiazole antibiotic that inhibits the synthesis of folic acid and thymine, and selectively suppresses Photobacterium phosphoreum bioluminescence. It demonstrates antibacterial activity against Escherichia coli-mediated urinary tract infections, but not against sulII gene-positive Escherichia coli. This product is applicable for research on urinary tract infections and Photobacterium phosphoreum bioluminescence mechanisms.
  • Serves as a reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable for HPLC, GC, and MS
Catalog No. 50-004-50573 Supplier Medchemexpress LLC Supplier No. 25MGHYB0333R
Sulfamethizole (Standard) is the analytical standard of Sulfamethizole (HY-B0333). It is intended for research and analytical applications. Sulfamethizole is a sulfathiazole antibiotic that inhibits the synthesis of folic acid and thymine, and selectively suppresses Photobacterium phosphoreum bioluminescence. It demonstrates antibacterial activity against Escherichia coli-mediated urinary tract infections, but not against sulII gene-positive Escherichia coli. This product is applicable for research on urinary tract infections and Photobacterium phosphoreum bioluminescence mechanisms.
  • Serves as a reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable for HPLC, GC, and MS