Learn More
Alkali Scientific Sulfamethoxazole, 100 Grams
SDP
Catalog No. 5000661248
Encompass_Preferred
Sulfamethoxazole is an antibiotic available in a 100g powder form, widely used to treat various bacterial infections. It is often combined with trimethoprim to form the co-trimoxazole combination, which enhances its effectiveness against a broad range of Gram-positive and Gram-negative bacteria. Sulfamethoxazole works by inhibiting bacterial folic acid synthesis, making it effective for treating urinary tract infections, respiratory infections, and gastrointestinal infections. This 100g size is typically used for large-scale preparations in clinical settings, laboratory research, and pharmaceutical manufacturing, offering a cost-effective solution for repeated use in diverse infection treatments.
Catalog No. 50-006-61248 Supplier Alkali Scientific Supplier No. SUL001CP
Sulfamethoxazole is an antibiotic available in a 100g powder form, widely used to treat various bacterial infections. It is often combined with trimethoprim to form the co-trimoxazole combination, which enhances its effectiveness against a broad range of Gram-positive and Gram-negative bacteria. Sulfamethoxazole works by inhibiting bacterial folic acid synthesis, making it effective for treating urinary tract infections, respiratory infections, and gastrointestinal infections. This 100g size is typically used for large-scale preparations in clinical settings, laboratory research, and pharmaceutical manufacturing, offering a cost-effective solution for repeated use in diverse infection treatments.