Learn More
Medchemexpress LLC Sulfamethoxazole-13C6 | 1196157-90-0 | 99.9% | 1 MG
SDP
Catalog No. 5000405417
Encompass_Preferred
Sulfamethoxazole-13C6 is a 13C labeled Sulfamethoxazole (Ro 4-2130-13C6), which is a sulfonamide antibiotic with widespread antibacterial activity. It inhibits bacterial folate metabolism by competing with 4-Aminobenzoic acid (PABA) to act on dihydropteroate synthetase and dihydropteroate reductase.
  • Suitable for the study of urinary tract infections (UTIs), prostatitis, and bronchitis.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-05417 Supplier Medchemexpress LLC Supplier No. HYB0322S11MG
Sulfamethoxazole-13C6 is a 13C labeled Sulfamethoxazole (Ro 4-2130-13C6), which is a sulfonamide antibiotic with widespread antibacterial activity. It inhibits bacterial folate metabolism by competing with 4-Aminobenzoic acid (PABA) to act on dihydropteroate synthetase and dihydropteroate reductase.
  • Suitable for the study of urinary tract infections (UTIs), prostatitis, and bronchitis.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.